The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026369.2 | NP_080645.2 | 2 - 3 | 400 | 87 |
GenBank mRNA | AK011348.1 | - | 2 - 3 | 277 | 87 |
AK011911.1 | - | 2 - 3 | 277 | 87 | |
AK017669.1 | - | 2 - 3 | 278 | 87 | |
AK017985.1 | - | 2 - 3 | 311 | 87 | |
AK019149.1 | - | 2 - 3 | 277 | 87 | |
AK087427.1 | - | 2 - 3 | 267 | 87 | |
AK149756.1 | - | 2 - 3 | 269 | 87 | |
AK150443.1 | - | 2 - 3 | 277 | 87 | |
AK150703.1 | - | 2 - 3 | 277 | 87 | |
AK151654.1 | - | 2 - 3 | 268 | 87 | |
AK151656.1 | - | 2 - 3 | 290 | 87 | |
AK152281.1 | - | 2 - 3 | 290 | 87 | |
AK153262.1 | - | 2 - 3 | 271 | 87 | |
AK159407.1 | - | 2 - 3 | 324 | 87 | |
AK159438.1 | - | 2 - 3 | 264 | 87 | |
AK159738.1 | - | 2 - 3 | 287 | 87 | |
AK166284.1 | - | 2 - 3 | 292 | 87 | |
AK166335.1 | - | 2 - 3 | 307 | 87 | |
AK166725.1 | - | 2 - 3 | 291 | 87 | |
AK166919.1 | - | 2 - 3 | 293 | 87 | |
AK167916.1 | - | 2 - 3 | 261 | 87 | |
AK168264.1 | - | 2 - 3 | 268 | 87 | |
AK191774.1 | - | 2 - 3 | 119 | 87 | |
AK208221.1 | - | 1 - 2 | 67 | 87 | |
AK209531.1 | - | 2 - 3 | 113 | 87 | |
BC060143.1 | - | 2 - 3 | 306 | 87 | |
CF170085.1 | - | 2 - 3 | 400 | 87 |