The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026850.4 | NP_081126.2 | 5 - 6 | 611 | 126 |
GenBank mRNA | AF110512.1 | - | 4 - 5 | 581 | 126 |
AK004334.1 | - | 5 - 6 | 606 | 126 | |
AK078372.1 | - | 5 - 6 | 612 | 126 | |
AK168509.1 | - | 5 - 6 | 652 | 126 | |
AK169572.1 | - | 5 - 6 | 602 | 126 | |
BC005601.1 | - | 5 - 6 | 632 | 126 |