Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nup37
Entrez Gene ID
69736
Gene Name
nucleoporin 37
Gene Aliases
2410003L22Rik, 2810039M17Rik
Chromosome Location
Chr.10: 88146976 - 88178395 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027191.2 NP_081467.2 7 - 8 864 98
NM_028334.4 NP_082610.1 7 - 8 882 98
GenBank mRNA AK010370.1 - 7 - 8 808 98
AK012890.1 - 7 - 8 878 98
AK040463.1 - 7 - 8 865 98
AK165537.1 - 7 - 8 828 98
BC011102.1 - 7 - 8 848 98
BC032180.1 - 6 - 7 792 98
BE951864.1 - 3 - 4 98

View Full Product Details