Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr20rt
Entrez Gene ID
70948
Gene Name
WD repeat domain 20, retrogene
Gene Aliases
4921538B03Rik, 4930427E19Rik, Wdr20, Wdr20b
Chromosome Location
Chr.12: 65225517 - 65228454 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027614.1 NP_081890.1 1 - 2 1983 109
GenBank mRNA AK015014.1 - 1 - 2 1983 109
AK015210.1 - - 1433 109
BC050778.1 - 1 - 2 1770 109

View Full Product Details