Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bloc1s2
Entrez Gene ID
73689
Gene Name
biogenesis of lysosomal organelles complex-1, subunit 2
Gene Aliases
2410089B13Rik, BLOS2, Bloc1s2a
Chromosome Location
Chr.19: 44139246 - 44146470 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028607.1 NP_082883.1 4 - 5 431 83
GenBank mRNA AK010745.1 - 4 - 5 428 83
AK145313.1 - 4 - 5 84
BC048500.1 - 3 - 4 311 83
BC065806.1 - 4 - 5 424 83

View Full Product Details