Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Commd9
Entrez Gene ID
76501
Gene Name
COMM domain containing 9
Gene Aliases
1810029F08Rik, AA409293
Chromosome Location
Chr.2: 101886262 - 101901639 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029635.3 NP_083911.1 5 - 6 467 109
GenBank mRNA AK007640.1 - 5 - 6 471 109
AK154731.1 - 5 - 6 473 109
BC030374.1 - 5 - 6 463 109
BC040688.1 - 5 - 6 441 109
BC043684.1 - 5 - 6 441 109
BC056201.1 - 4 - 5 402 109
BC060058.1 - 5 - 6 465 109

View Full Product Details