Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrgprh
Entrez Gene ID
80978
Gene Name
MAS-related GPR, member H
Gene Aliases
Gpr90, Mgrb, MrgH
Chromosome Location
Chr.17: 12876034 - 12877842 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030726.1 NP_109651.1 1 - 1 1646 68
GenBank mRNA AF295365.1 - 1 - 1 1646 68
BC116696.1 - 1 - 1 1276 68
BC117525.1 - 1 - 1 1274 68

View Full Product Details