Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btnl2
Entrez Gene ID
547431
Gene Name
butyrophilin-like 2
Gene Aliases
BTL-II, BTLN2, Btl2, Gm315, NG9
Chromosome Location
Chr.17: 34354789 - 34369492 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079835.2 NP_524574.1 7 - 8 1609 83
GenBank mRNA AK136521.1 - 7 - 8 1611 83
BC141103.1 - 7 - 8 1499 83

View Full Product Details