The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133228.3 | NP_573491.2 | 3 - 4 | 377 | 92 |
GenBank mRNA | AK030724.1 | - | 3 - 4 | 324 | 92 |
AK049711.1 | - | 3 - 4 | 296 | 92 | |
BC031988.1 | - | 3 - 4 | 280 | 92 |