Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tifab
Entrez Gene ID
212937
Gene Name
TRAF-interacting protein with forkhead-associated domain, family member B
Gene Aliases
-
Chromosome Location
Chr.13: 56173702 - 56179018 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145976.4 NP_666088.1 1 - 2 341 95
GenBank mRNA AK036643.1 - 1 - 2 258 95

View Full Product Details