The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013707.2 | NP_038735.2 | 1 - 1 | 449 | 77 |
GenBank mRNA | AF003691.1 | - | 1 - 1 | 438 | 77 |
BC104262.1 | - | 1 - 1 | 423 | 77 | |
BC104263.1 | - | 1 - 1 | 422 | 77 | |
D85925.1 | - | 1 - 1 | 438 | 77 |