Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B9d1
Entrez Gene ID
27078
Gene Name
B9 protein domain 1
Gene Aliases
AW045994, B9, Eppb9
Chromosome Location
Chr.11: 61505168 - 61512929 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013717.2 NP_038745.1 4 - 5 380 114
GenBank mRNA AB030483.1 - 4 - 5 380 114
AK003038.1 - 4 - 5 375 114
AK010355.1 - 4 - 5 409 114
AK131701.1 - 4 - 5 371 114
BC008113.1 - 4 - 5 348 114

View Full Product Details