Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sfrp4
Entrez Gene ID
20379
Gene Name
secreted frizzled-related protein 4
Gene Aliases
-
Chromosome Location
Chr.13: 19623175 - 19632825 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016687.3 NP_057896.1 1 - 2 709 103
GenBank mRNA AF117709.1 - 1 - 2 724 103
AK080766.1 - 1 - 2 710 103
BC034853.1 - 1 - 2 668 103

View Full Product Details