Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gzmn
Entrez Gene ID
245839
Gene Name
granzyme N
Gene Aliases
GrN
Chromosome Location
Chr.14: 56165786 - 56174612 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB049454.2 - 5 - 6 702 93
BC120809.1 - 4 - 5 623 93

View Full Product Details