The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172155.1 | NP_742167.1 | 3 - 4 | 331 | 103 |
GenBank mRNA | AF525160.1 | - | 3 - 4 | 331 | 103 |
BC064735.1 | - | 3 - 4 | 337 | 102 | |
BC119340.1 | - | 3 - 4 | 325 | 103 | |
BC119344.1 | - | 3 - 4 | 325 | 103 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172156.2 | NP_742168.1 | - | 331 | 103 |
GenBank mRNA | AF525161.1 | - | - | 331 | 103 |
BC068253.1 | - | - | 328 | 103 |