The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172277.2 | NP_758481.1 | 10 - 11 | 1278 | 89 |
GenBank mRNA | AK045061.1 | - | 10 - 11 | 1271 | 89 |
AK082871.1 | - | 10 - 11 | 1271 | 89 | |
AK148429.1 | - | 10 - 11 | 1271 | 89 | |
AK156817.1 | - | 10 - 11 | 1224 | 89 | |
AK161672.1 | - | 9 - 10 | 1212 | 89 | |
BC019142.1 | - | 9 - 10 | 1274 | 89 | |
BC037599.1 | - | 10 - 11 | 1304 | 89 |