The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172624.3 | NP_766212.2 | 19 - 20 | 2618 | 103 |
GenBank mRNA | AK050021.1 | - | 19 - 20 | 2560 | 103 |
AK050023.1 | - | 19 - 20 | 2620 | 103 | |
AK131178.1 | - | 17 - 18 | 4601 | 103 | |
BC057631.1 | - | 19 - 20 | 2617 | 103 |