Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpp9
Entrez Gene ID
224897
Gene Name
dipeptidylpeptidase 9
Gene Aliases
6430584G11Rik, A330078I11, DPP IX, DPRP2
Chromosome Location
Chr.17: 56186674 - 56218889 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172624.3 NP_766212.2 7 - 8 1029 102
GenBank mRNA AK039652.1 - 6 - 7 1122 102
AK050021.1 - 7 - 8 971 102
AK050023.1 - 7 - 8 1031 102
AK078301.1 - 7 - 8 977 102
AK131178.1 - 4 - 5 2892 102
AK219044.1 - 1 - 2 46 102
BC057631.1 - 7 - 8 1028 102

View Full Product Details