Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpp9
Entrez Gene ID
224897
Gene Name
dipeptidylpeptidase 9
Gene Aliases
6430584G11Rik, A330078I11, DPP IX, DPRP2
Chromosome Location
Chr.17: 56186674 - 56218889 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172624.3 NP_766212.2 8 - 9 1156 85
GenBank mRNA AK039652.1 - 7 - 8 1249 85
AK050021.1 - 8 - 9 1098 85
AK050023.1 - 8 - 9 1158 85
AK078301.1 - 8 - 9 1104 85
AK131178.1 - 5 - 6 3019 85
BC057631.1 - 8 - 9 1155 85

View Full Product Details