The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173347.2 | NP_775482.1 | 3 - 4 | 794 | 80 |
GenBank mRNA | AK081637.1 | - | 3 - 4 | 485 | 80 |
AK142160.1 | - | 3 - 4 | 393 | 80 | |
BC044877.1 | - | 1 - 2 | 219 | 80 | |
BC057546.1 | - | 3 - 4 | 794 | 80 | |
BC058635.1 | - | 3 - 4 | 523 | 80 |