Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hist1h2ah
- Entrez Gene ID
-
319168
- Gene Name
- histone cluster 1, H2ah
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 22035122 - 22035643 on Build GRCm38
- Gene Symbol
- Hist1h2ac
- Entrez Gene ID
-
319164
- Gene Name
- histone cluster 1, H2ac
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 23683473 - 23683959 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178189.5 |
NP_835496.1 |
1 - 1 |
87 |
88 |
- Gene Symbol
- Hist1h2ag
- Entrez Gene ID
-
319167
- Gene Name
- histone cluster 1, H2ag
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 22042478 - 22042949 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178186.3 |
NP_835493.1 |
1 - 1 |
67 |
88 |
GenBank mRNA |
BC065803.1 |
- |
1 - 1 |
67 |
88 |
- Gene Symbol
- Hist1h2ai
- Entrez Gene ID
-
319191
- Gene Name
- histone cluster 1, H2ai
- Gene Aliases
-
H2a-291A
- Chromosome Location
-
Chr.13: 21716412 - 21716859 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC116373.2 |
- |
1 - 1 |
45 |
88 |
- Gene Symbol
- Hist1h2an
- Entrez Gene ID
-
319170
- Gene Name
- histone cluster 1, H2an
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 21786772 - 21787218 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178184.2 |
NP_835491.1 |
1 - 1 |
45 |
88 |
View Full Product Details