Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt15
Entrez Gene ID
319508
Gene Name
synaptotagmin XV
Gene Aliases
CHR10SYT, E230025K04Rik, sytXV
Chromosome Location
Chr.14: 34220046 - 34230421 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176931.2 NP_795905.2 4 - 5 765 81
NM_181529.4 NP_852682.1 4 - 5 765 81
GenBank mRNA AB109019.1 - 3 - 4 637 81
AB109020.1 - 3 - 4 81
AK042641.1 - 4 - 5 81
AK054180.1 - 4 - 5 765 81
BC139313.1 - 4 - 5 676 81
BC139314.1 - 4 - 5 677 81

View Full Product Details