The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195284.1 | NP_001182213.1 | 1 - 2 | 142 | 96 |
NM_176966.4 | NP_795940.1 | 1 - 2 | 142 | 96 | |
GenBank mRNA | AK038893.1 | - | 1 - 2 | 120 | 96 |
AK140020.1 | - | 1 - 2 | 126 | 96 | |
BC130265.1 | - | 1 - 2 | 58 | 96 | |
CJ115006.1 | - | 1 - 2 | 142 | 96 |