The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177820.4 | NP_808488.2 | 2 - 3 | 207 | 102 |
GenBank mRNA | AK033678.1 | - | - | 179 | 102 |
AK144113.1 | - | 2 - 3 | 210 | 102 | |
BC147348.1 | - | - | 153 | 102 | |
BC147349.1 | - | - | 153 | 102 |