Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bpi
Entrez Gene ID
329547
Gene Name
bactericidal permeablility increasing protein
Gene Aliases
9230105K17Rik, Bpifd1
Chromosome Location
Chr.2: 158258091 - 158284533 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177850.3 NP_808518.1 2 - 3 296 82
GenBank mRNA AK033770.1 - 2 - 3 298 82
AY363993.1 - 2 - 3 294 82
AY648037.1 - 2 - 3 296 82
AY853179.1 - 2 - 3 281 82
BC118049.1 - 2 - 3 290 82
BC118504.1 - 2 - 3 290 82

View Full Product Details