The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178799.3 | NP_848914.2 | 2 - 3 | 369 | 90 |
GenBank mRNA | AK033963.1 | - | 2 - 3 | 288 | 90 |
AK080793.1 | - | 2 - 3 | 369 | 90 | |
BB643798.1 | - | 2 - 3 | 369 | 90 |