Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbed4
Entrez Gene ID
223773
Gene Name
zinc finger, BED type containing 4
Gene Aliases
1700009F06Rik
Chromosome Location
Chr.15: 88751666 - 88784517 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181412.3 NP_852077.1 1 - 2 94 97
GenBank mRNA CD350815.1 - 1 - 2 81 97

View Full Product Details