The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182694.2 | NP_874353.2 | 1 - 2 | 233 | 105 |
GenBank mRNA | BC089358.1 | - | 1 - 2 | 233 | 105 |