Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oog2
Entrez Gene ID
381570
Gene Name
oogenesin 2
Gene Aliases
C85069
Chromosome Location
Chr.4: 144190717 - 144196934 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198661.3 NP_941063.1 1 - 2 76 85
GenBank mRNA AK139660.1 - 1 - 2 78 85

View Full Product Details