Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ssxb5
- Entrez Gene ID
-
387586
- Gene Name
- synovial sarcoma, X member B, breakpoint 5
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 8803691 - 8809445 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199319.3 |
NP_955023.1 |
6 - 7 |
556 |
110 |
- Gene Symbol
- Ssxb1
- Entrez Gene ID
-
67985
- Gene Name
- synovial sarcoma, X member B, breakpoint 1
- Gene Aliases
-
4930414C09Rik
- Chromosome Location
-
Chr.X: 8413305 - 8422157 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC171964.1 |
- |
- |
|
110 |
- Gene Symbol
- Ssxb10
- Entrez Gene ID
-
385312
- Gene Name
- synovial sarcoma, X member B, breakpoint 10
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 8327424 - 8336235 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199064.1 |
NP_951019.1 |
7 - 8 |
|
110 |
GenBank mRNA |
AY244801.1 |
- |
7 - 8 |
|
110 |
View Full Product Details