Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pgk1
Entrez Gene ID
18655
Gene Name
phosphoglycerate kinase 1
Gene Aliases
Pgk-1
Chromosome Location
Chr.X: 106187100 - 106203699 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008828.3 NP_032854.2 1 - 1 75 102
GenBank mRNA BY292706.1 - 1 - 1 76 102

View Full Product Details