The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009391.3 | NP_033417.1 | 1 - 1 | 50 | 74 |
GenBank mRNA | AK002436.1 | - | 1 - 1 | 141 | 74 |