The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033593.3 | NP_291071.1 | 1 - 1 | 1545 | 99 |
GenBank mRNA | AK043330.1 | - | - | 99 | |
AY013793.1 | - | 1 - 1 | 1545 | 99 | |
BC035540.1 | - | 1 - 1 | 1700 | 99 |