The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145405.2 | NP_663380.1 | 4 - 4 | 574 | 96 |
NR_045779.1 | 3 - 3 | 468 | 96 | ||
GenBank mRNA | AK048689.1 | - | 3 - 3 | 411 | 96 |
BC010817.1 | - | 4 - 4 | 553 | 96 | |
BM932729.1 | - | 4 - 4 | 548 | 96 |