The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177744.4 | NP_808412.1 | 4 - 4 | 2944 | 82 |
GenBank mRNA | AK033521.1 | - | 4 - 4 | 2945 | 82 |