The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177748.2 | NP_808416.1 | 9 - 9 | 1650 | 120 |
GenBank mRNA | AF548540.1 | - | 9 - 9 | 1650 | 120 |