Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btbd3
Entrez Gene ID
228662
Gene Name
BTB (POZ) domain containing 3
Gene Aliases
mKIAA0952
Chromosome Location
Chr.2: 138256584 - 138287424 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025431.1 NP_001020602.1 1 - 1 54 93
GenBank mRNA BY012054.1 - 1 - 1 74 93

View Full Product Details