Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Lingo2
Entrez Gene ID
242384
Gene Name
leucine rich repeat and Ig domain containing 2
Gene Aliases
B230217C06Rik, Lern3, Lrrn6c
Chromosome Location
Chr.4: 35706647 - 36952254 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166000.1 NP_001159472.1 2 - 2 148 101
GenBank mRNA BC090619.1 - 2 - 2 76 101

View Full Product Details