Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jmjd4
Entrez Gene ID
194952
Gene Name
jumonji domain containing 4
Gene Aliases
6430559I23
Chromosome Location
Chr.11: 59450045 - 59458567 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_037992.1 3 - 4 768 75
GenBank mRNA AK032477.1 - 3 - 4 742 75
AK038650.1 - 2 - 3 311 75
AK039116.1 - 3 - 4 446 75
AK047586.1 - 3 - 4 769 75
AK171919.1 - 3 - 4 723 75
BC049148.1 - 3 - 4 753 75
BC118020.1 - 3 - 4 698 75


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB632454.1 - 2 - 3 311 Chr. 11 - 59450664 - 59455075 [+] on Build GRCm38 75
GenBankmRNA BB632949.1 - 3 - 4 442 Chr. 11 - 59450371 - 59454985 [+] on Build GRCm38 75


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream synaptosomal-associated protein, 47 Snap47 1110031B06Rik, SNAP-47 -3.5kb Anti-sense Chr. - 59407141 - 59450029 on Build GRCm38
Downstream zinc finger protein 867 Zfp867 7.1kb Anti-sense Chr. - 59461197 - 59472566 on Build GRCm38

View Full Product Details