The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205068.1 | NP_001191997.1 | 4 - 5 | 1006 | 66 |
NM_178659.6 | NP_848774.1 | 4 - 5 | 1042 | 66 | |
NR_037992.1 | 4 - 5 | 956 | 66 | ||
GenBank mRNA | AK032477.1 | - | 4 - 5 | 930 | 66 |
AK037954.1 | - | 4 - 5 | 1041 | 66 | |
AK038650.1 | - | 3 - 4 | 499 | 66 | |
AK039116.1 | - | 4 - 5 | 634 | 66 | |
AK047586.1 | - | 4 - 5 | 957 | 66 | |
AK052144.1 | - | 4 - 5 | 992 | 66 | |
AK079104.1 | - | 4 - 5 | 1000 | 66 | |
AK084669.1 | - | 4 - 5 | 1001 | 66 | |
AK145394.1 | - | 4 - 5 | 1020 | 66 | |
AK171919.1 | - | 4 - 5 | 911 | 66 | |
BC049148.1 | - | 4 - 5 | 941 | 66 | |
BC098198.1 | - | 3 - 4 | 3599 | 66 | |
BC116707.1 | - | 4 - 5 | 973 | 66 | |
BC118020.1 | - | 4 - 5 | 886 | 66 |