Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snord8
Entrez Gene ID
100217445
Gene Name
small nucleolar RNA, C/D box 8
Gene Aliases
MBII-28
Chromosome Location
Chr.14: 52209786 - 52209884 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028542.1 1 - 1 44 91
GenBank mRNA AF357382.1 - 1 - 1 44 91


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream suppressor of Ty 16 Supt16 Cdc68, Fact140, Spt16, Supt16h -12.5kb Sense Chr. - 52160419 - 52197239 on Build GRCm38
Downstream RAB2B, member RAS oncogene family Rab2b 1500012D09Rik, 4930528G15Rik, A230002G14, D530043M21Rik 51.9kb Sense Chr. - 52261759 - 52279395 on Build GRCm38

View Full Product Details