Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir7-1
- Entrez Gene ID
-
723902
- Gene Name
- microRNA 7-1
- Gene Aliases
-
Mirn7-1, mir-7a-1, mmu-mir-7-1
- Chromosome Location
-
Chr.13: 58392779 - 58392886 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029825.1 |
|
1 - 1 |
55 |
93 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA881786.1 |
- |
1 |
106 |
|
Chr. 13 - 58392382 - 58392937 [-] on Build GRCm38 |
93 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
heterogeneous nuclear ribonucleoprotein K |
Hnrnpk |
Hnrpk, KBBP, NOVA |
|
Sense |
Chr. - 58391132 - 58403343 on Build GRCm38 |
Downstream |
RMI1, RecQ mediated genome instability 1, homolog (S. cerevisiae) |
Rmi1 |
4932432N11Rik, C79893 |
9.4kb
|
Anti-sense |
Chr. - 58402248 - 58411149 on Build GRCm38 |
View Full Product Details