Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt74
Entrez Gene ID
406222
Gene Name
keratin 74
Gene Aliases
K74, Kb37
Chromosome Location
Chr.15: 101754259 - 101763504 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033444.1 8 - 9 1297 76


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream keratin 71 Krt71 AA589543, Ca, Cal4, Cu, Krt2-6g, mK6irs, mK6irs1 -11.2kb Sense Chr. - 101733949 - 101743097 on Build GRCm38
Downstream keratin 72 Krt72 AI507495, K6irs2, Krt2-35, Krt72-ps 21.4kb Sense Chr. - 101776560 - 101786458 on Build GRCm38

View Full Product Details