Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gm10636
- Entrez Gene ID
-
100038607
- Gene Name
- predicted gene 10636
- Gene Aliases
-
ENSMUSG00000074175
- Chromosome Location
-
Chr.3: 146367153 - 146378944 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033542.1 |
|
1 - 2 |
225 |
92 |
GenBank mRNA |
AK135478.1 |
- |
1 - 2 |
226 |
92 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CJ051954.1 |
- |
1 - 2 |
226 |
|
Chr. 3 - 146368886 - 146378944 [-] on Build GRCm38 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
lysophosphatidic acid receptor 3 |
Lpar3 |
Edg7, lpA3 |
-91.2kb
|
Anti-sense |
Chr. - 146220961 - 146286214 on Build GRCm38 |
Downstream |
synovial sarcoma, X breakpoint 2 interacting protein |
Ssx2ip |
AU014939, AU042321, Adip |
25.7kb
|
Anti-sense |
Chr. - 146404617 - 146440144 on Build GRCm38 |
View Full Product Details