Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Olfr75-ps1
- Entrez Gene ID
-
258186
- Gene Name
- olfactory receptor 75, pseudogene 1
- Gene Aliases
-
MOR135-20, Olfr75, V1
- Chromosome Location
-
Chr.11: 73405450 - 73409858 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002859.2 |
|
1 - 2 |
110 |
103 |
GenBank mRNA |
DR065618.1 |
- |
1 - 2 |
110 |
103 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DR065921.1 |
- |
|
111 |
|
Chr. - - [-] on Build |
103 |
GenBankmRNA |
DR065662.1 |
- |
1 - 2 |
113 |
|
Chr. 11 - 73405747 - 73409826 [-] on Build GRCm38 |
103 |
GenBankmRNA |
CB173622.1 |
- |
1 - 2 |
74 |
|
Chr. 11 - 73405895 - 73409787 [-] on Build GRCm38 |
103 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
olfactory receptor 1 |
Olfr1 |
I54, MOR135-13 |
-6.0kb
|
Sense |
Chr. - 73395076 - 73399495 on Build GRCm38 |
Downstream |
olfactory receptor 378 |
Olfr378 |
MOR135-2 |
15.2kb
|
Sense |
Chr. - 73425037 - 73428477 on Build GRCm38 |
View Full Product Details