Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Scarna3b
- Entrez Gene ID
-
100217447
- Gene Name
- small Cajal body-specific RNA 3B
- Gene Aliases
-
MBI-100
- Chromosome Location
-
Chr.12: 80391614 - 80391737 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_028544.1 |
|
1 - 1 |
69 |
105 |
GenBank mRNA |
AF357403.1 |
- |
1 - 1 |
69 |
105 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 1843a |
Mir1843a |
Mir1843, mir-1843a, mmu-mir-1843, mmu-mir-1843a |
|
Sense |
Chr. - 80391613 - 80391677 on Build GRCm38 |
Downstream |
predicted gene, 30025 |
Gm30025 |
|
43.2kb
|
Anti-sense |
Chr. - 80434951 - 80435938 on Build GRCm38 |
View Full Product Details