The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177775.1 | NP_001171246.1 | 5 - 6 | 821 | 92 |
GenBank mRNA | AB284505.1 | - | - | 898 | 92 |