Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc42b
Entrez Gene ID
546886
Gene Name
coiled-coil domain containing 42B
Gene Aliases
Gm5988
Chromosome Location
Chr.5: 120628335 - 120634235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195094.1 NP_001182023.1 4 - 5 461 80

View Full Product Details