Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gc
Entrez Gene ID
14473
Gene Name
group specific component
Gene Aliases
DBP, VDB
Chromosome Location
Chr.5: 89417511 - 89457898 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008096.2 NP_032122.1 1 - 2 237 88
GenBank mRNA AK010965.1 - 1 - 2 61 88
BC010762.1 - 1 - 2 100 88
BC051395.1 - 1 - 2 104 88
M55413.1 - 1 - 2 47 88

View Full Product Details