Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gc
Entrez Gene ID
14473
Gene Name
group specific component
Gene Aliases
DBP, VDB
Chromosome Location
Chr.5: 89417511 - 89457898 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008096.2 NP_032122.1 9 - 10 1343 98
GenBank mRNA AK010965.1 - 9 - 10 1167 98
BC010762.1 - 9 - 10 1206 98
BC051395.1 - 9 - 10 1210 98
M55413.1 - 9 - 10 1153 98

View Full Product Details